1-Fluoro-4-iodobenzene ≥98.0% stabilised
Leverantör: TCI
Varning
Synonyms: 4-Fluoroiodobenzene
Stabilization: Stabilised with copper chip
F0237-25G F0237-5G
TCIAF0237-25GEA 535 SEK
TCIAF0237-25G TCIAF0237-5G
1-Fluoro-4-iodobenzene ≥98.0% stabilised
1-Fluoro-4-iodobenzene
Formel: C₆H₄FI
Kokpunkt: 183°C
Smältpunkt: –18 °C
Densitet: 1,9253 g/cm³ (15 °C)
Flampunkt: 68°C
MDL Number: MFCD00001052
CAS nummer: 352-34-1
EINECS: 206-522-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdfc36d0c8db0cf142c5a90f082c6d2575d29ea0&vid=b9f34a370c474dc6e7fcd5a01a05ed93c06f6506&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %