3'-Trifluoromethoxyacetophenone ≥97.0%

Leverantör: TCI
Varning

Synonyms: 3'-(Trifluoromethoxy)acetophenone, 1-[3-(Trifluoromethoxy)phenyl]-1-ethanone, 1-Ethanoyl-3-(trifluormethoxy)benzol

T1587-1G T1587-5G
TCIAT1587-1GEA 230 SEK
TCIAT1587-1G TCIAT1587-5G
3'-Trifluoromethoxyacetophenone ≥97.0%
3'-Trifluoromethoxyacetophenone
Formel: C₉H₇F₃O₂
Flampunkt: 83°C
MDL Number: MFCD00042403
CAS nummer: 170141-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f236a7b32680e28229bf57d685ea3da7452ca75&vid=4f236a7b32680e28229bf57d685ea3da7452ca75&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR