n-Hentriacontane ≥95.0%
Leverantör: TCI
H0012-1G
H0012-10G
TCIAH0012-1GEA
1100
SEK
TCIAH0012-1G
TCIAH0012-10G
n-Hentriacontane ≥95.0%
n-Hentriacontane
Formel:
H₃C(CH₂)₂₉CH₃ MW: 436.85 g/mol Kokpunkt: 180°C Storage Temperature: Omgivande |
MDL Number:
MFCD00048745 CAS nummer: 630-04-6 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 66.0 to 70.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...