1-Jodpropan ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Propyl iodide
Stabilization: Stabilised with copper chip
I0068-25G I0068-500G I0068-100G
TCIAI0068-25GEA 775 SEK
TCIAI0068-25G TCIAI0068-500G TCIAI0068-100G
1-Jodpropan ≥98.0% stabilised
1-Jodpropan
Formel: C₃H₇I
MW: 169,99 g/mol
Kokpunkt: 102°C
Smältpunkt: –98 °C
Densitet: 1,741 g/cm³ (20 °C)
Flampunkt: 44°C
Storage Temperature: Omgivande
MDL Number: MFCD00001095
CAS nummer: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 14,07862
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2355784bd416f7570ee096c5fbb9eb84620cb24f&vid=33df6068363349f90fc01f705038506bcf671ab7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %