Chloroacetaldehyde dimethyl acetal ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Chloro-2,2-dimethoxyethane
C0085-25ML C0085-500ML
TCIAC0085-25MLEA 215 SEK
TCIAC0085-25ML TCIAC0085-500ML
Chloroacetaldehyde dimethyl acetal ≥98.0%
Chloroacetaldehyde dimethyl acetal
Formel: C₄H₉ClO₂
MW: 124,57 g/mol
Kokpunkt: 130°C
Smältpunkt: < –73 °C
Densitet: 1,098 g/cm³ (20 °C)
Flampunkt: 35°C
Storage Temperature: Omgivande
MDL Number: MFCD00000948
CAS nummer: 97-97-2
EINECS: 202-624-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3456a638f937acc34d8950c7ca993bd8e12efd5&vid=e3c6928f09dbdee1f57eee7558fe9e92d08a95d7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %