Tritylium tetrafluoroborate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Triphenylcarbenium tetrafluoroborate

T1173-25G T1173-5G
TCIAT1173-25GEA 1425 SEK
TCIAT1173-25G TCIAT1173-5G
Tritylium tetrafluoroborate ≥98.0%
Tritylium tetrafluoroborate
Formel: C₁₉H₁₅BF₄
MW: 330,13 g/mol
Smältpunkt: 202°C
Storage Temperature: Kylskåp
MDL Number: MFCD00013120
CAS nummer: 341-02-6
EINECS: 206-433-3
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01169755771416068e819a95600ea410bb82d8bd&vid=01169755771416068e819a95600ea410bb82d8bd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR