Tetrabutylammonium tetrahydroborate ≥96.0% reducing reagent

Leverantör: TCI
Fara

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

T0917-25G T0917-5G
TCIAT0917-25GEA 1600 SEK
TCIAT0917-25G TCIAT0917-5G
Tetrabutylammonium tetrahydroborate ≥96.0% reducing reagent
Tetrabutylammonium tetrahydroborate
Formel: C₁₆H₄₀BN
MW: 257.31 g/mol
Smältpunkt: 126°C
MDL Number: MFCD00012035
CAS nummer: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5e42f005e030ef2df50693950dcd5fb352f0c60&vid=48afed30cb3ed0307ec7ebe8868010fca582e2f9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR