Monobenzon ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-(Fenylmetoxi)fenol

B1104-25G B1104-500G B1104-100G
TCIAB1104-25GEA 350 SEK
TCIAB1104-25G TCIAB1104-500G TCIAB1104-100G
Monobenzon ≥99.0%
Monobenzon
Formel: C₁₃H₁₂O₂
MW: 200,24 g/mol
Kokpunkt: 328 °C (1013 hPa)
Smältpunkt: 117,9…121.5 °C
Densitet: 1,26 g/cm³ (20 °C)
MDL Number: MFCD00002333
CAS nummer: 103-16-2
EINECS: 203-083-3
Merck Index: 14,06248
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a6ab75fca7252429e61470f19592d362e786610&vid=1a6ab75fca7252429e61470f19592d362e786610&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 120.0 to 124.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR