Iridium(III) acetylacetonate

Leverantör: TCI
Varning

Synonyms: Iridium(III) 2,4-acetylacetonate, Tris(2,4-pentanedionato-O,O')iridium (III), Iridium (III) tris(2,4-pentanedionato-O,O'), Iridium (III) 2,4-pentanedionate

T2557-1G
TCIAT2557-1GEA 5350 SEK
TCIAT2557-1G
Iridium(III) acetylacetonate
Iridium(III) acetylacetonate
Formel: C₁₅H₂₁IrO₆
MW: 489.55 g/mol
MDL Number: MFCD00015353
CAS nummer: 15635-87-7
EINECS: 239-711-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26df3264c0c9dce9227c1c7e00c42c37c19933a9&vid=26df3264c0c9dce9227c1c7e00c42c37c19933a9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Elemental analysis (Carbon) 35.0 to 38.6 %
iridium 38.0 to 41.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR