Toluen-α-sulfonylfluorid ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride
B1062-25G B1062-5G
TCIAB1062-25GEA 1550 SEK
TCIAB1062-25G TCIAB1062-5G
Toluen-α-sulfonylfluorid ≥98.0%
Toluen-α-sulfonylfluorid
Formel: C₇H₇FO₂S
MW: 174,2 g/mol
Kokpunkt: 85°C
Smältpunkt: 91…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007424
CAS nummer: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III
Merck Index: 14,07542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=509a951303cdb2aca181f88f039db636cb5713ed&vid=509a951303cdb2aca181f88f039db636cb5713ed&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 95.0 deg-C