Styren ≥99.0% stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: Vinylbenzen

Stabilization: med 4-tert-Butylpyrokatekol

S0095-25ML S0095-500ML
TCIAS0095-25MLEA 215 SEK
TCIAS0095-25ML TCIAS0095-500ML
Styren ≥99.0% stabilised
Styren
Formel: C₈H₈
MW: 104,15 g/mol
Kokpunkt: 145°C
Smältpunkt: –31°C
Densitet: 0,906 g/cm³ (20 °C)
Flampunkt: 32°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008612
CAS nummer: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3,III
Merck Index: 14,08860
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f73ab98d681693e869739e38317e23d32abe467&vid=2f73ab98d681693e869739e38317e23d32abe467&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR