4'-Methylacetanilide ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: p-Acetotoluidid, p-Acetotoluidide, N-(4-Methylphenyl)acetamide
A0064-25G A0064-100G
TCIAA0064-25GEA 470 SEK
TCIAA0064-25G TCIAA0064-100G
4'-Methylacetanilide ≥98.0%
4'-Methylacetanilide
Formel: C₉H₁₁NO
MW: 149,19 g/mol
Kokpunkt: 307 °C (1013 hPa)
Smältpunkt: 145…148 °C
Densitet: 1,22 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008677
CAS nummer: 103-89-9
EINECS: 203-155-4
Merck Index: 13,00076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37bb0bc01bb89314fc28572d2bb255e578069ce8&vid=37bb0bc01bb89314fc28572d2bb255e578069ce8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 148.0 to 152.0 deg-C
Solubility in Methanol almost transparency