Adipohydrazide ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Adipyl hydrazide

A0170-25G A0170-250G
TCIAA0170-25GEA 230 SEK
TCIAA0170-25G TCIAA0170-250G
Adipohydrazide ≥99.0%
Adipohydrazide
Formel: C₆H₁₄N₄O₂
MW: 174,2 g/mol
Kokpunkt: 446 °C (1013 hPa)
Smältpunkt: 181…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007614
CAS nummer: 1071-93-8
EINECS: 213-999-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40f34fb6492f5e6c8d8062952ebc19781140088e&vid=40f34fb6492f5e6c8d8062952ebc19781140088e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Bromination Method) min. 98.0 %
Melting point 179.0 to 184.0 deg-C
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR