Tetrametylsilan ≥99.0% for NMR spectroscopy
Leverantör: TCI
Fara
T0154-100ML T0154-25ML T0154-500ML
TCIAT0154-100MLEA 1200 SEK
TCIAT0154-100ML TCIAT0154-25ML TCIAT0154-500ML
Tetrametylsilan ≥99.0% for NMR spectroscopy
Tetrametylsilan
Formel: C₄H₁₂Si
Kokpunkt: 26°C
Smältpunkt: –99 °C
Densitet: 0,642 g/cm³ (20 °C)
Flampunkt: –27°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008274
CAS nummer: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
Merck Index: 14,09229
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2d6f43af8f59a29a1b1afb3e58cb6c0a220c5ea&vid=0d7f6557940b3beed579411bc7c31c67f2a11433&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Specific gravity (20/20) 0.6380 to 0.6420
Refractive index n20/D 1.3570 to 1.3600