Methyl octanoate ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
O0033-25ML
O0033-500ML
TCIAO0033-25MLEA
310
SEK
TCIAO0033-25ML
TCIAO0033-500ML
Methyl octanoate ≥99.0%
Methyl octanoate
Formel:
C₉H₁₈O₂ MW: 158,24 g/mol Kokpunkt: 193°C Smältpunkt: –36,7 °C Densitet: 0,877 g/cm³ (20 °C) Flampunkt: 76°C |
MDL Number:
MFCD00009551 CAS nummer: 111-11-5 EINECS: 203-835-0 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...