L(-)-Xylose ≥97.0%

Leverantör: TCI EUROPE

Synonyms: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

X0021-25G X0021-5G
TCIAX0021-25GEA 1750 SEK
TCIAX0021-25G TCIAX0021-5G
L(-)-Xylose ≥97.0%
L(-)-Xylose
Formel: C₅H₁₀O₅
MW: 150,13 g/mol
Kokpunkt: 331 °C (1013 hPa)
Smältpunkt: 150…152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151096
CAS nummer: 609-06-3
EINECS: 210-174-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f0a8e4c602b29642f22c6d6f460df06ebfa90b3&vid=9f0a8e4c602b29642f22c6d6f460df06ebfa90b3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Periodic acid Method) min. 97.0 %
Specific rotation [a]20/D -18.0 to -21.0 deg (C=10, H2O)
Solubility in Water within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR