L(+)-Fructose ≥95.0%

Leverantör: TCI

Synonyms: L(+)-Levulose

F0317-100MG F0317-1G
TCIAF0317-100MGEA 1250 SEK
TCIAF0317-100MG TCIAF0317-1G
L(+)-Fructose ≥95.0%
L(+)-Fructose
Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05662378
CAS nummer: 7776-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25c900cb884e45f12620f75a04ef875f57c8efff&vid=25c900cb884e45f12620f75a04ef875f57c8efff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 95.0 area%
Specific rotation [a]20/D +84.0 to +94.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR