(±)-2-Methyl-1-butanol ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
M0175-25ML
M0175-500ML
TCIAM0175-25MLEA
255
SEK
TCIAM0175-25ML
TCIAM0175-500ML
(±)-2-Methyl-1-butanol ≥97.0%
(±)-2-Methyl-1-butanol
Formel:
C₅H₁₂O MW: 88,15 g/mol Kokpunkt: 128°C Smältpunkt: –70 °C Densitet: 0,82 g/cm³ (20 °C) Flampunkt: 43°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004743 CAS nummer: 137-32-6 EINECS: 205-289-9 UN: 1105 ADR: 3,III Merck Index: 14,06030 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...