Etylendiamin, vattenfri ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 1,2-Diaminoetan
E0077-25ML E0077-500ML
TCIAE0077-25MLEA 215 SEK
TCIAE0077-25ML TCIAE0077-500ML
Etylendiamin, vattenfri ≥98.0% (by GC, titration analysis)
Etylendiamin
Formel: C₂H₈N₂
MW: 60,1 g/mol
Kokpunkt: 116°C
Smältpunkt: 8,5 °C
Densitet: 0,898 g/cm³ (20 °C)
Flampunkt: 34°C
Storage Temperature: Omgivande
MDL Number: MFCD00008204
CAS nummer: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 14,03795
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a78a3d2881080e64b9f8dbac699424e45f606ec5&vid=3c52aee6bb207c4f1f21dffe598634973a08861f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %