2-(Methylvinyl)naphthalene ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 2-Isopropenylnaphthalene
I0413-25G I0413-5G
TCIAI0413-25GEA 1700 SEK
TCIAI0413-25G TCIAI0413-5G
2-(Methylvinyl)naphthalene ≥98.0%
2-(Methylvinyl)naphthalene
Formel: C₁₃H₁₂
MW: 168,24 g/mol
Kokpunkt: 155°C
Storage Temperature: Kylskåp
MDL Number: MFCD00191524
CAS nummer: 3710-23-4
EINECS: 223-053-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c5e87dba1b4c630068a468c2c89337fba93fc1c&vid=6c5e87dba1b4c630068a468c2c89337fba93fc1c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 55.0 deg-C