Titanium(IV) butanolate
Leverantör: TCI EUROPE
Varning
Synonyms: Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide
B0742-25G B0742-500G
TCIAB0742-25GEA 245 SEK
TCIAB0742-25G TCIAB0742-500G
Titanium(IV) butanolate
Titanium(IV) butanolate
Formel: C₁₆H₃₆O₄Ti
MW: 340,34 g/mol
Kokpunkt: 310 °C (1013 hPa)
Smältpunkt: –55 °C
Densitet: 0,993 g/cm³ (20 °C)
Flampunkt: 46°C
MDL Number: MFCD00009433
CAS nummer: 5593-70-4
EINECS: 227-006-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51643979ff74e929fd24a23769e2908f61470110&vid=51643979ff74e929fd24a23769e2908f61470110&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Ignition residue 23.2 to 24.5 %