Indol ≥99.0%
Leverantör: TCI
Fara
Synonyms: 1H-Indole
I0021-25G I0021-100G
TCIAI0021-25GEA 245 SEK
TCIAI0021-25G TCIAI0021-100G
Indol ≥99.0%
Indole
Formel: C₈H₇N
Kokpunkt: 253°C
Smältpunkt: 50…51 °C
Densitet: 1,22 g/cm³ (20 °C)
Flampunkt: 110 °C
MDL Number: MFCD00005607
CAS nummer: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,04963
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d0985c6da19418f498fb4b4e83f5103cc828dbe&vid=5d0985c6da19418f498fb4b4e83f5103cc828dbe&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 51.0 to 54.0 deg-C
Solubility in EtOH almost transparency