Vinylphosphonic acid ≥95.0%

Leverantör: TCI
Fara

V0068-25G V0068-250G
TCIAV0068-25GEA 870 SEK
TCIAV0068-25G TCIAV0068-250G
Vinylphosphonic acid ≥95.0%
Vinylphosphonic acid
Formel: C₂H₅O₃P
MDL Number: MFCD00043866
CAS nummer: 1746-03-8
EINECS: 217-123-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55fb8b125dd1193c4143ade1fec3f72bedf0f4a9&vid=55fb8b125dd1193c4143ade1fec3f72bedf0f4a9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (Bromination Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR