(R)-(-)-α-Cyclohexanemethylamine 98%, ee 94%, ChiPros®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (R)-(-)-1-Cyclohexylethylamine, (R)-1-Cyclohexylethylamine
L19051.03 L19051.06
L19051.03EA 560 SEK
L19051.03 L19051.06
(R)-(-)-α-Cyclohexanemethylamine 98%, ee 94%, ChiPros®
(R)-(-)-α-Cyclohexanemethylamine
Purity: 98% CAS No.: 5913-13-3 Molecular Formula: C8H17N Formula Weight: 127.23 Flash Point: 51°(123°F) Refractive Index: 1.4620 Odour: Amine-like pH: 11 Boiling Point: 177-178° Beilstein Reference: 2935070 Optical Rotation: -4° (neat) UN No.: 2733 MDL No.: MFCD00043338
Formel: C₈H₁₇N
MW: 127.23 g/mol
Kokpunkt: 177…178 °C
Densitet: 0.866
Flampunkt: 51 °C (123 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00043338
CAS nummer: 5913-13-3
EINECS: 227-634-2
UN: 2733
ADR: 3 III