(S)-(-)-3-Phenylcyclohexanone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (S)-3-Phenylcyclohexanone

P1500-1G P1500-100MG
TCIAP1500-1GEA 9225 SEK
TCIAP1500-1G TCIAP1500-100MG
(S)-(-)-3-Phenylcyclohexanone ≥98.0%
(S)-(-)-3-Phenylcyclohexanone
Formel: C₁₂H₁₄O
MW: 174,2390 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD08276411
CAS nummer: 57344-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e8cc1b5756a0994d48d9ac2566abbe563c3854&vid=a9e8cc1b5756a0994d48d9ac2566abbe563c3854&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 95.0 ee%
Specific rotation [a]20/D -18.0 to -22.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR