(R)-(+)-3-Phenylcyclohexanone ≥97.0%
Leverantör: TCI EUROPE
P1499-1G
P1499-100MG
TCIAP1499-1GEA
8550
SEK
TCIAP1499-1G
TCIAP1499-100MG
(R)-(+)-3-Phenylcyclohexanone ≥97.0%
(R)-(+)-3-Phenylcyclohexanone
Formel:
C₁₂H₁₄O Storage Temperature: Kylskåp |
CAS nummer:
34993-51-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Optical purity (LC) | min. 95.0 ee% |
Specific rotation [a]20/D | +10.0 to +12.0 deg (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...