(±)-1,1'-Bi-2-naphthol ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: 1,1'-Binaphthyl-2,2'-Diol, 2,2'-Dihydroxy-1,1'-binaphthyl, 1,1'-Bi-2-naphthol, [1,1'-Binaphthalene]-2,2'-diol
A11182.06 A11182.22
A11182.06EA 970 SEK
A11182.06 A11182.22
(±)-1,1'-Bi-2-naphthol ≥99%
(±)-1,1'-Bi-2-naphthol
Purity: 99% CAS No.: 602-09-5 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odour: Characteristic Melting Point: 215-218° Beilstein Reference: 997518 Merck Reference: 14,1226 UN No.: 2811 MDL No.: MFCD00004068
Formel: C₂₀H₁₄O₂
MW: 286,33 g/mol
Smältpunkt: 215…218 °C
Densitet: 1,301
Storage Temperature: Omgivande
MDL Number: MFCD00004068
CAS nummer: 602-09-5
EINECS: 210-014-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,1226