1,3-Dihydroxyacetone dimer ≥96.0%

Leverantör: TCI
Varning

Synonyms: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol

D0560-25G D0560-500G D0560-100G
TCIAD0560-25GEA 215 SEK
TCIAD0560-25G TCIAD0560-500G TCIAD0560-100G
1,3-Dihydroxyacetone dimer ≥96.0%
1,3-Dihydroxyacetone dimer
Formel: C₆H₁₂O₆
Storage Temperature: Kylskåp
MDL Number: MFCD00051019
CAS nummer: 26776-70-5
Merck Index: 14,03182
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ea4340e07e39fdc81c759d54c4c1e0b3eed5e5a&vid=7ea4340e07e39fdc81c759d54c4c1e0b3eed5e5a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Periodic acid Method) min. 96.0 %
Solubility in Water very faint turbidity
Water max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR