Methyl-6-fluoro-2-naphthoate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 6-Fluoro-2-naphthoic acid methyll ester

F0473-1G F0473-5G
TCIAF0473-1GEA 2400 SEK
TCIAF0473-1G TCIAF0473-5G
Methyl-6-fluoro-2-naphthoate ≥98.0%
Methyl-6-fluoro-2-naphthoate
Formel: C₁₂H₉FO₂
MDL Number: MFCD06797106
CAS nummer: 5043-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426514fead14ba98579755dcb97248e8cd6544fa&vid=426514fead14ba98579755dcb97248e8cd6544fa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR