Methyl-2-naphthoate ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: Methyl 2-naphthoate
L09973.03 L09973.06
L09973.03EA 615 SEK
L09973.03 L09973.06
Methyl-2-naphthoate ≥99%
Methyl-2-naphthoate
Purity: 99% CAS No.: 2459-25-8 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 75-79° Boiling Point: 290° MDL No.: MFCD00017345
Formel: C₁₂H₁₀O₂
Kokpunkt: 290 °C
Smältpunkt: 75…79 °C
Storage Temperature: Omgivande
MDL Number: MFCD00017345
CAS nummer: 2459-25-8