6-Bromo-2-naphthoic acid ≥98%
Leverantör: Thermo Scientific
Varning
L16093.06 L16093.14
L16093.06EA 1150 SEK
L16093.06 L16093.14
6-Bromo-2-naphthoic acid ≥98%
6-Bromo-2-naphthoic acid
Purity: 98% CAS No.: 5773-80-8 Molecular Formula: C11H7BrO2 Formula Weight: 251.08 Melting Point: 292-296° Beilstein Reference: 2328940
Formel: C₁₁H₇BrO₂
MW: 251,08 g/mol
Smältpunkt: 292…296 °C
Storage Temperature: Omgivande
MDL Number: MFCD01075720
CAS nummer: 5773-80-8