4-Nitrostyrene ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Nitro-4-vinylbenzene, 1-Ethenyl-4-nitrobenzene
Stabilization: med 4-tert-Butylpyrokatekol
N0503-1G N0503-5G
TCIAN0503-1GEA 845 SEK
TCIAN0503-1G TCIAN0503-5G
4-Nitrostyrene ≥98.0% stabilised
4-Nitrostyrene
Formel: C₈H₇NO₂
MW: 149,15 g/mol
Storage Temperature: Frys
MDL Number: MFCD00041254
CAS nummer: 100-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=546166bb5f86b4a22dba14cf28e11abf24c0896d&vid=546166bb5f86b4a22dba14cf28e11abf24c0896d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow powder to lump
Purity (GC) min. 98.0 %