4-(Trifluoromethylthio)benzaldehyde ≥90%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-((Trifluoromethyl)thio)benzaldehyde
B20313.03 B20313.06
B20313.03EA 1225 SEK
B20313.03 B20313.06
4-(Trifluoromethylthio)benzaldehyde ≥90%
4-(Trifluoromethylthio)benzaldehyde
Purity: 90+% CAS No.: 4021-50-5 Molecular Formula: C8H5F3OS Formula Weight: 206.19 Flash Point: 96°(204°F) Refractive Index: 1.5110 Odour: Stench Melting Point: 29-31° Boiling Point: 219° Beilstein Reference: 2328411 MDL No.: MFCD00129191
Formel: C₈H₅F₃OS
MW: 206.19 g/mol
Kokpunkt: 219 °C
Smältpunkt: 29…31 °C
Densitet: 1.310
Flampunkt: 96 °C (204 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00129191
CAS nummer: 4021-50-5