Methyl-2-amino-5-chlorobenzoate ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 2-amino-5-chlorobenzoate
M1877-1G
M1877-5G
TCIAM1877-1GEA
215
SEK
TCIAM1877-1G
TCIAM1877-5G
Methyl-2-amino-5-chlorobenzoate ≥98.0% (by GC, titration analysis)
Methyl-2-amino-5-chlorobenzoate
Formel:
C₈H₈ClNO₂ MW: 185,61 g/mol Kokpunkt: 170°C |
MDL Number:
MFCD00007837 CAS nummer: 5202-89-1 EINECS: 225-992-4 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...