Ethyl-3-aminocrotonate ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Ethyl 3-aminobut-2-enoate
A12991.14 A12991.36
A12991.14EA 625 SEK
A12991.14 A12991.36
Ethyl-3-aminocrotonate ≥98%
Ethyl-3-aminocrotonate
Purity: 98+% CAS No.: 626-34-6 Molecular Formula: C6H11NO2 Formula Weight: 129.16 Flash Point: 97°(206°F) Refractive Index: 1.4990 Melting Point: 29-35° Boiling Point: 210-215° Beilstein Reference: 471284 UN No.: 3263 MDL No.: MFCD00008073
Formel: C₆H₁₁NO₂
MW: 129,16 g/mol
Kokpunkt: 210…215 °C
Smältpunkt: 29…35 °C
Densitet: 1,022
Flampunkt: 97 °C (206 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008073
CAS nummer: 7318-00-5
EINECS: 230-782-0
ADR: 8,II