Methyl-4-bromocrotonate ≥80.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Methyl 4-bromobut-2-enoate
M1689-25G M1689-5G
TCIAM1689-25GEA 950 SEK
TCIAM1689-25G TCIAM1689-5G
Methyl-4-bromocrotonate ≥80.0%
Methyl-4-bromocrotonate
Formel: C₅H₇BrO₂
MW: 179,01 g/mol
Kokpunkt: 75°C
Flampunkt: 92°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000246
CAS nummer: 1117-71-1
EINECS: 214-251-0
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9629b5a6eb7b7e25ad80e8f11e21ce2802c93c3d&vid=9629b5a6eb7b7e25ad80e8f11e21ce2802c93c3d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Brown clear liquid
Purity (GC) min. 80.0 %