4-Dibenzofuranboronic acid ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L19830.03
L19830.06
L19830.03EA
790
SEK
L19830.03
L19830.06
4-Dibenzofuranboronic acid ≥98%
Purity: 98+%
CAS No.: 100124-06-9
Molecular Formula: C12H9BO3
Formula Weight: 212.01
Melting Point: 286-291°
Beilstein Reference: 1645426
Formel:
C₁₂H₉BO₃ Smältpunkt: 286…291 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00092336 CAS nummer: 100124-06-9 |