Ferrocenemethanol 95,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: Hydroxymethylferrocene

H0464-1G H0464-5G
TCIAH0464-1GEA 935 SEK
TCIAH0464-1G TCIAH0464-5G
Ferrocenemethanol 95,0 GC_ANALYSMETOD
Ferrocenemethanol
Formel: C₁₁H₁₂FeO
MW: 216.06 g/mol
MDL Number: MFCD00040878
CAS nummer: 1273-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a57478077afc36c72c3ed130209c98c2a828fef&vid=0a57478077afc36c72c3ed130209c98c2a828fef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 95.0 %
Melting point 75.0 to 80.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR