(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0%

Leverantör: TCI
Varning

D2528-1G D2528-200MG
TCIAD2528-1GEA 2450 SEK
TCIAD2528-1G TCIAD2528-200MG
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0%
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
Formel: C₁₄H₁₉FeN
MW: 257.16 g/mol
Kokpunkt: 93°C
MDL Number: MFCD00066273
CAS nummer: 31886-58-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69127a98c9d04a237114ce9fb84cc81b2854537e&vid=69127a98c9d04a237114ce9fb84cc81b2854537e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Orange to Brown clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR