(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%
Leverantör: Thermo Fisher Scientific
Varning
L19679.03 L19679.MD
L19679.03EA 6550 SEK
L19679.03 L19679.MD
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine
Purity: 97% CAS No.: 31886-58-5 Molecular Formula: C14H19FeN Formula Weight: 257.16 Flash Point: 110°(230°F) Refractive Index: 1.5890 Boiling Point: 120-121°/0.7mm Optical Rotation: +13° (c=1 in ethanol, 5H) MDL No.: MFCD00066273
Formel: C₁₄H₁₉FeN
MW: 257.16 g/mol
Kokpunkt: 120…121 °C (0.7 mmHg)
Densitet: 1.222
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00066273
CAS nummer: 31886-58-5