(±)-3-Aminoquinuclidine dihydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

A1236-25G A1236-5G
TCIAA1236-25GEA 4450 SEK
TCIAA1236-25G TCIAA1236-5G
(±)-3-Aminoquinuclidine dihydrochloride ≥98.0%
(±)-3-Aminoquinuclidine dihydrochloride
Formel: C₇H₁₆Cl₂N₂
MW: 199,1213 g/mol
MDL Number: MFCD00137395
CAS nummer: 6530-09-2
EINECS: 229-424-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0574e72f5289c12733951b8368038acfe2914382&vid=0574e72f5289c12733951b8368038acfe2914382&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR