Quinuclidine hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Azabicyclo[2.2.2]octane hydrochloride

Q0050-5G Q0050-1G
TCIAQ0050-5GEA 6750 SEK
TCIAQ0050-5G TCIAQ0050-1G
Quinuclidine hydrochloride ≥98.0%
Quinuclidine hydrochloride
Formel: C₇H₁₄ClN
MW: 147,650 g/mol
Smältpunkt: 365°C
MDL Number: MFCD00012727
CAS nummer: 39896-06-5
EINECS: 254-682-1
Merck Index: 14,08081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d90eb0d487ed2a74780c20f1e7f2f205d0be8887&vid=d90eb0d487ed2a74780c20f1e7f2f205d0be8887&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR