3,4-(Methylenedioxy)phenylacetic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: (1,3-Benzodioxol-5-yl)acetic acid
A11229.06 A11229.14 A11229.22
A11229.06EA 1400 SEK
A11229.06 A11229.14 A11229.22
3,4-(Methylenedioxy)phenylacetic acid ≥98%
3,4-(Methylenedioxy)phenylacetic acid
Purity: 98% CAS No.: 2861-28-1 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 127-130° Beilstein Reference: 177739
Formel: C₉H₈O₄
MW: 180.16 g/mol
Smältpunkt: 127…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014576
CAS nummer: 2861-28-1
EINECS: 220-679-9