3,4-(Methylenedioxy)phenylacetonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M1923-25G
M1923-5G
TCIAM1923-25GEA
3550
SEK
TCIAM1923-25G
TCIAM1923-5G
3,4-(Methylenedioxy)phenylacetonitrile ≥98.0%
3,4-(Methylenedioxy)phenylacetonitrile
Formel:
C₉H₇NO₂ MW: 161,16 g/mol Kokpunkt: 140°C |
MDL Number:
MFCD00005835 CAS nummer: 4439-02-5 EINECS: 224-655-9 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...