2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid
B23610.03 B23610.06
B23610.03EA 1950 SEK
B23610.03 B23610.06
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid ≥97%
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid
Purity: 97+% CAS No.: 656-46-2 Molecular Formula: C8H4F2O4 Formula Weight: 202.12 Melting Point: 154-157° MDL No.: MFCD00792417
Formel: C₈H₄F₂O₄
MW: 202,12 g/mol
Smältpunkt: 154…157 °C
Storage Temperature: Omgivande
MDL Number: MFCD00792417
CAS nummer: 656-46-2