2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-5-carbaldehyde
B24232.03 B24232.06
B24232.03EA 1425 SEK
B24232.03 B24232.06
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde ≥97%
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde
Purity: 97% CAS No.: 656-42-8 Molecular Formula: C8H4F2O3 Formula Weight: 186.11 Refractive Index: 1.4990 Boiling Point: 103-105°/20mm MDL No.: MFCD00792420
Formel: C₈H₄F₂O₃
MW: 186,11 g/mol
Kokpunkt: 103…105 °C (20 mmHg)
Densitet: 1,399
Storage Temperature: Omgivande
MDL Number: MFCD00792420
CAS nummer: 656-42-8