Piperonylonitrile ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,4-(Methylenedioxy)benzonitrile, 1,3-Benzodioxole-5-carbonitrile
P1074-10G
P1074-25G
TCIAP1074-10GEA
575
SEK
TCIAP1074-10G
TCIAP1074-25G
Piperonylonitrile ≥98.0%
Piperonylonitrile
Formel:
C₈H₅NO₂ MW: 147,13 g/mol |
MDL Number:
MFCD00005820 CAS nummer: 4421-09-4 EINECS: 224-590-6 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Light red to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 95.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...