5-Methoxypiperonal ≥97%
Leverantör: Thermo Scientific
Synonyms: 7-Methoxybenzo[d][1,3]dioxole-5-carbaldehyde
L08213.06 L08213.14 L08213.03
L08213.06EA 3725 SEK
L08213.06 L08213.14 L08213.03
5-Methoxypiperonal ≥97%
5-Methoxypiperonal
Purity: 97% CAS No.: 5780-07-4 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 126-132° Beilstein Reference: 384077
Formel: C₉H₈O₄
MW: 180,16 g/mol
Smältpunkt: 126…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016900
CAS nummer: 5780-07-4
EINECS: 227-304-8