Tetraethyl methylenebisphosphonate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Tetraethyl methylenediphosphonate, Tetraethyl methylenebis(phosphonate)
A14532.06 A14532.14 A14532.22
A14532.06EA 1800 SEK
A14532.06 A14532.14 A14532.22
Tetraethyl methylenebisphosphonate ≥98%
Tetraethyl methylenebisphosphonate
Purity: 98+% CAS No.: 1660-94-2 Molecular Formula: C9H22O6P2 Formula Weight: 288.22 Flash Point: 110°(230°F) Refractive Index: 1.4400 Boiling Point: 151-153°/1.2mm Beilstein Reference: 1813241 MDL No.: MFCD00039887
Formel: C₉H₂₂O₆P₂
MW: 288,22 g/mol
Kokpunkt: 151…153 °C (1,2 mmHg)
Densitet: 1,162
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039887
CAS nummer: 1660-94-2
EINECS: 216-764-5