Diethyl (hydroxymethyl)phosphonate 97%
Leverantör: Thermo Scientific
Varning
320950050 320950250
ACRO320950050EA 1150 SEK
ACRO320950050 ACRO320950250
Diethyl (hydroxymethyl)phosphonate 97%
Diethyl (hydroxymethyl)phosphonate
Formel: C₅H₁₃O₄P
MW: 168,13 g/mol
Kokpunkt: 124…126 °C (3 mmHg)
Densitet: 1,14 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014418
CAS nummer: 3084-40-0
EINECS: 221-391-6
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %